ContactUsLinkComponent
Eisco® Fractures, Faults, and Dislocations Model
Catalogue # 470353-778
Fournisseur:  Wards
undefined
Eisco® Fractures, Faults, and Dislocations Model
Catalogue # 470353-778
Fournisseur:  Wards
Supplier Number:  BD0080
Spécifications
  • Height:
    10"
  • Length:
    22"
  • Width:
    10"
Spécifications
À propos de cet article
Geology model illustrating fault types and stress effects on rock layers for hands-on learning experiences.
  • Durable model made from high-quality polymer
  • Eco-friendly; hand painted with non-toxic paint
  • Hands-on learning aid
This rock layer model is an essential tool for geology education, designed to help students visualize how rock formations respond to stress. By showcasing different fault types (normal, reverse, and strike-slip), it provides an interactive way to understand geological forces at play. The vivid colors and distinct fault line divisions aid in clearly differentiating the hanging wall from the footwall, making it easier for students to grasp complex geological concepts.
Ideal for hands-on demonstrations, the model allows educators to illustrate the dynamic nature of the Earth’s crust. It serves as a versatile teaching aid, beneficial for both introductory geology lessons and more advanced discussions on tectonic activity. Whether used in a classroom, lab, or educational exhibit, this model enhances comprehension and engagement, making abstract geological concepts more tangible and accessible.