DURAN® Original GL 25 Laboratory Bottle Screw Cap, with Lip Seal, PP, Blue
Catalogue # 77623-798
Fournisseur: DWK Life Sciences (KIMBLE)
New Product
Spécifications
- Ø:33 mm
- Color:Blue
- Height:19 mm
- Material:PP
Spécifications
À propos de cet article
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colors: Blue, green, yellow and gray with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Cap with integral lip seal provides effective sealing for short term storage
- Pouring ring for drip-free pouring
-
The color helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
Autoclavable
Fully compatible with the GL 45 threads and therefore highly versatile.