Order Entry
Canada
ContactUsLinkComponent
Eisco® Friction Board Apparatus
Catalog # 470345-756
Supplier:  Wards
undefined
Eisco® Friction Board Apparatus
Catalog # 470345-756
Supplier:  Wards
Supplier Number:  PH0336

Specifications

  • Height:
    0.75"
  • Length:
    19"
  • Width:
    3"

Specifications

About this item

Hands-on tool for comparative friction studies.

  • Includes two sliders with different surface textures

  • Precision-crafted wooden base with metal plane

This apparatus provides a practical way to observe how surface materials influence resistance to motion. Suitable for use across educational and research environments, it supports repeatable experiments in mechanics and force analysis.