ContactUsLinkComponent
Cleanroom Wipes and Bottle Holder
  77898-360
New Product
undefined
Cleanroom Wipes and Bottle Holder
  77898-360
 :  GD-0306
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Wall mounted wipes and bottle holder
  • W×D×H:
    16×4.5×12"
Palbam’s wipes and bottle holder is a stainless steel, wall-mountable solution designed for cleanrooms. They ensure organized, contamination-free storage of wipes and bottles, promoting efficiency, accessibility, and compliance in controlled environments.
  • Stainless steel 304 with electropolished finish
  • Holds wipes and IPA bottles
The gownroom is often the first impression for employees and visitors alike. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all ergonomic, cleanliness, and functional requirements.