Cleanroom Utility Carts
Fournisseur: PALBAM 2008 AC LTD TE
New Product
Certificats
À propos de cet article
The cleanroom carts are designed to withstand heavy loads. Up to 60 kg per shelf. Integral raised ledges prevent parts from rolling off and are easy to wipe down.
304 stainless steel
- Electropolished
- White nylon 5" cleanroom casters (two with brakes) supplied with every cart
The gown room is often the first impression for employees and visitors alike, that they are entering into a special area. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all the ergonomic, cleanliness and functional requirements.
Ordering information:
Options include two or three shelves, solid or perforated, contact your Avantor representative for more information.
Frequently Bought Together