About this item
Labcon's ZAP™ SLIK low retention aerosol filter pipette tips have been proven in case studies to block aerosol contamination and reduce fluid retention. Featuring a pure 10μm pore size patented filter without additives, they keep pipettor barrels clean of DNA, radionuclides, and other contaminants. The contaminant-free filter ensures sample integrity even upon contact. Each ZAP™ SLIK tip is precisely designed to fit the range of pipettors listed in Labcon online fit charts. Each fitting is meticulously inspected to ensure proper fit without leakage or the need for extra force.
Patented 10 µm, hydrophobic filter and hydrophobic resin minimize sample retention and ensure contamination-free pipetting
- Uniquely engineered to fit most popular brands of pipettes
- Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
- Tip sizes range from 10µl to 1000µl
- Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
- Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
- Made in USA
Note: Low retention tips are best suited for pipetting viscous samples, proteins, and DNA, but may not be appropriate for all uses. Labcon resins are pre-tested for metals contamination.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates. View tip compatibility guides online at labcon.com/tipfits.
Endotoxin-free, ATP-free, RNase/DNase-free, Low Trace Metals (<1 ppm), Protease-free, DNA-free.