Eclipse™ Standard Surface Pipette Tips
À propos de cet article
Labcon's Eclipse™ standard surface pipette tips are made of high-quality, 100% pure virgin medical grade polypropylene. The resin is pre-tested to ensure no extractables are present.
Uniquely engineered to fit most popular brands of pipettes
- Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
- Tip sizes range from 10µl to 1250µl
- Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
- Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
- Made in USA
Labcon resins are pre-tested to ensure no extractables, PFAS, or other compounds are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates. View tip compatibility guides online at labcon.com/tipfits.
Certifications: PFAS-free, Endotoxin-free, ATP-free, RNase/Dnase-free, Low Trace Metals (<1 ppm).
14 Options Available Below
Product Details & Documents
Labcon's Eclipse™ standard surface pipette tips are made of high-quality, 100% pure virgin medical grade polypropylene. The resin is pre-tested to ensure no extractables are present.
Uniquely engineered to fit most popular brands of pipettes
- Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
- Tip sizes range from 10µl to 1250µl
- Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
- Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
- Made in USA
Labcon resins are pre-tested to ensure no extractables, PFAS, or other compounds are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates. View tip compatibility guides online at labcon.com/tipfits.
Certifications: PFAS-free, Endotoxin-free, ATP-free, RNase/Dnase-free, Low Trace Metals (<1 ppm).
Product Family Options
Refill Eclipse™ tips
Informations sur le produit
- Tip StyleVolumeSterileLongueurTip ColorFor Use WithPackedAvailabilityPrice
- Graduated, UltraFine10 µl-31.4 mmClearUniversal pipettes1248/pack, 12480/case, 10 packs/caseLog in pour la disponibilitéPack of 1,248Pack of 1,248 - $71.87$71.87Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 12480Case of 12480 - $718.60$718.60Prix promotionnel pour les nouveaux clientsPack of 1,248$71.87Case of 12480$718.60Supplier Number: 1038-260-000-9
- Bevel, Graduated200 µl-49.7 mmClearUniversal pipettes960/pack, 9600/case, 10 packs/caseLog in pour la disponibilitéPack of 960Pack of 960 - $34.17$34.17Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 9600Case of 9600 - $341.68$341.68Prix promotionnel pour les nouveaux clientsPack of 960$34.17Case of 9600$341.68Supplier Number: 1093-260-000-9
- Bevel200 µl-49.7 mmJaune/clairUniversal pipettes960/pack, 9600/case, 10 packs/caseLog in pour la disponibilitéPack of 960Pack of 960 - $32.58$32.58Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 9600Case of 9600 - $325.84$325.84Prix promotionnel pour les nouveaux clientsPack of 960$32.58Case of 9600$325.84Supplier Number: 1030-260-000-9
Racked Eclipse™ tips
Informations sur le produit
- Tip StyleVolumeSterileLongueurTip ColorFor Use WithPackedAvailabilityPrice
- Bevel200 µl-49.7 mmJaune/clairUniversal pipettes960/pack, 9600/case, 10 packs/caseLog in pour la disponibilitéPack of 960Pack of 960 - $54.22$54.22Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 9600Case of 9600 - $542.02$542.02Prix promotionnel pour les nouveaux clientsPack of 960$54.22Case of 9600$542.02Supplier Number: 1030-960-008-9
- Bevel200 µl+49.7 mmJaune/clairUniversal pipettes960/pack, 9600/case, 10 packs/casePack of 960$80.67Case of 9600$806.70Supplier Number: 1030-965-008-9
- Graduated, UltraFine, Extra Long10 µl-47.6 mmClearUniversal pipettes960/pack, 9600/case, 10 packs/caseLog in pour la disponibilitéPack of 960Pack of 960 - $84.78$84.78Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 9600Case of 9600 - $847.76$847.76Prix promotionnel pour les nouveaux clientsPack of 960$84.78Case of 9600$847.76Supplier Number: 1037-960-008-9
- Graduated, UltraFine, Extra Long10 µl+47.6 mmClearUniversal pipettes960/pack, 9600/case, 10 packs/casePack of 960$108.35Case of 9600$1,083.50Supplier Number: 1037-965-008-9
- Graduated, UltraFine10 µl-31.4 mmClearUniversal pipettes1152/pack, 11520/case, 10 packs/casePack of 1,152$93.42Case of 11520$934.20Supplier Number: 1038-960-038-9
- Graduated, UltraFine10 µl+31.4 mmClearUniversal pipettes1152/pack, 11520/case, 10 packs/casePack of 1,152$102.68Case of 11520$1,026.83Supplier Number: 1038-965-038-9
- Bevel, Graduated200 µl-49.7 mmClearUniversal pipettes960/pack, 9600/case, 10 packs/caseLog in pour la disponibilitéPack of 960Pack of 960 - $60.50$60.50Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 9600Case of 9600 - $605.04$605.04Prix promotionnel pour les nouveaux clientsPack of 960$60.50Case of 9600$605.04Supplier Number: 1093-960-008-9
- Bevel, Graduated200 µl+49.7 mmClearUniversal pipettes960/pack, 9600/case, 10 packs/casePack of 960$77.36Case of 9600$773.49Supplier Number: 1093-965-008-9
Refill Eclipse™ FlexTop™ tips
Informations sur le produit
- Tip StyleVolumeSterileLongueurTip ColorFor Use WithPackedAvailabilityPrice
- Extended, Flextop, UltraFine1250 µl-88.9 mmClearUniversal pipettes480/pack, 4800/case, 10 packs/caseLog in pour la disponibilitéPack of 480Pack of 480 - $38.31$38.31Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 4800Case of 4800 - $383.12$383.12Prix promotionnel pour les nouveaux clientsPack of 480$38.31Case of 4800$383.12Supplier Number: 1045-260-000-9
Racked Eclipse™ FlexTop™ tips
Informations sur le produit
- Tip StyleVolumeSterileLongueurTip ColorFor Use WithPackedAvailabilityPrice
- Extended, Flextop, UltraFine1250 µl-88.9 mmClearUniversal pipettes576/pack, 4608/case, 8 packs/caseLog in pour la disponibilitéPack of 576Pack of 576 - $67.66$67.66Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 4608Case of 4608 - $541.25$541.25Prix promotionnel pour les nouveaux clientsPack of 576$67.66Case of 4608$541.25Supplier Number: 1045-960-008-9
- Extended, Flextop, UltraFine1250 µl+88.9 mmClearUniversal pipettes576/pack, 4608/case, 8 packs/casePack of 576$89.25Case of 4608$713.99Supplier Number: 1045-965-008-9