Eclipse™ Pipette Tips for Rainin LTS Pipettes
: Labcon
New Product
Labcon tips for LTS pipettes are made from special flexible resins. For easiest ejection apply less than 3 kg of force when mounting. Do not force tips onto the pipette barrel with excessive force.
Engineered to fit old and new Rainin LTS pipettes
- Ergonomic, flexible top eliminates excessive mounting force
- Ultrafine point ensures precision and accuracy in sample delivery
- Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
- Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
- Made in USA
Labcon resins are pre-tested to ensure no extractables are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates.
:
All tips are Endotoxin-free, ATP-free, RNase/Dnase-free, Low Trace Metals (<1 ppm). Select tips are Protease-free, DNA-free, PFAS-free.
Frequently Bought Together