À propos de cet article
High quality combined with excellent design, the Palbam Class waste receptacles are an ideal addition to any cleanroom. Choose from three options: free stand on casters, floor mount on leg stand, or wall mount.
All bins include an insert designed to hold the bin liner in place
- Large lid openings are designed to ensure minimum contact with users
- In stock fabrication material: 304 stainless steels, electropolished
- Immediate delivery
The gown room is often the first impression for employees and visitors alike. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all the ergonomic, cleanliness, and functional requirements.
These round waste receptacles include an internal nylon holder and lid. The lid has a large opening for easy disposal. No need to come in contact with the bin (no flaps or other devices are attached, as these just increase contact between user and the waste bin). The free stand model is mounted on casters which are hidden from view. It can also be mounted on a bench or to the wall.