Vikan® UST Tank Brushes | Avantor
Vikan® UST Tank Brushes
Supplier:   Remco Products
Vikan® UST Tank Brushes
Supplier:  Remco Products
Certificates
Vikan® UST Tank Brushes
Vikan® UST Tank Brushes
8 Options Available Below
Product Details & Documents
Designed to incorporate Vikan®’s Ultra Safe Technology (UST), this industrial tank brush offers unparalleled bristle security and hygiene.
  • Angled bristles facilitate optimal cleaning
  • Wide drainage holes prevent water pooling and aid drying during storage
  • Threaded connection
  • Overall dimensions: 6.7W×4.1H×7.5"L
This UST tank scrub offers optimal performance in scrubbing efficacy, durability, and hygienic design. The seamless, easy-to-clean design minimizes the risk of contamination, making it an ideal tool for tank cleaning in dairies, breweries, wineries, and other food and beverage production facilities. The UST tank brush is compact and lightweight, making it perfect for cleaning the insides of complex equipment like mixers and blenders. A perfect fit for any color-coded cleaning system.
Certifications: FDA compliant.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur totale
    Width
    Availability
    Price
  • UST tank brush, stiff bristles, pink
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70301
  • UST tank brush, stiff bristles, green
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70302
  • UST tank brush, stiff bristles, blue
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70303
  • UST tank brush, stiff bristles, red
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70304
  • UST tank brush, stiff bristles, white
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70305
  • UST tank brush, stiff bristles, yellow
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70306
  • UST tank brush, stiff bristles, orange
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70307
  • UST tank brush, stiff bristles, purple
    7.5"
    6.7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.36
    $148.36
    Each
       $148.36
    Supplier Number: 70308