Sorenson™ Universal TipStation Pipette Tips
Fournisseur: Sorenson Bioscience
Certificats
À propos de cet article
Universal Fit TipStation pipette tips are available sterile or non-sterile in 20 and 200 µl, and are a perfect platform for multi-channel pipetting.
Ideal platform for multi-channel pipettors or high throughput laboratories
- Can position lid to expose 96 or 192 tips
- Certified RNase/DNase free, human DNA free, and non-pyrogenic
- Wide compatibility across popular pipettors
Certifications:
ISO 13485, ISO 14001.
Frequently Bought Together