Sorenson™ HybridRack LTS Compatible Filtered Pipette Tips
: Corning
New Product
HybridRack LTS compatible filtered tips can help reduce plastic usage by 70% compared to traditional tip racks and are available sterile in volumes of 20, 200 and 1000 µl.
Compatible with LTS pipettors
Extended length options available in 20 and 1000 µl
Reduce plastic by 70% compared to traditional tip racks
Pipette directly from HybridRack with single or multi-channel pipettors
Certified RNase/DNase free, human DNA free, and non-pyrogenic
Filters feature 20 µm pore size
:
ISO 13485, ISO 14001.
Frequently Bought Together