Contrex® CF Liquid Detergent Concentrate
Catalog # CA75856-898
Supplier: Decon Labs
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
Specifications
About this item
Contrex® CF is a cation-free and phosphate-free liquid detergent concentrate.
Free-rinsing
- Phosphate-free
- Disposable straight to drain
Contrex® CF is formulated for use in cleaning of equipment and electronics that are sensitive to sodium and metal cations and also phosphates. Can be used for soak, scrub, sonication, and auto washers.