À propos de cet article
PROCLEAR polypropylene filters are designed for a wide range of prefiltration duties within the production of pharmaceuticals. They are particularly suited for applications where chemical compatibility is an issue.
Graded density polypropylene media for high capacity
- Extremely robust to withstand aggressive conditions
- All polypropylene construction
- MURUS and DEMICAPs can be gamma-irradiated and autoclaved
- PFAS-free options available
The optimum pleat configuration and graded density polypropylene media used in PROCLEAR PP filters ensure the filters have the highest possible throughput to blockage, resulting in long service life.
The PROCLEAR PP range of filters are fully supported by a comprehensive validation guide to simplify its specification into new and existing processes.
Additional connection configurations are available.
DEMI Capsules are supplied three units per pack. FB Extension stand for filling bell included.