Saturix Disposable Heavy-Weight Microfiber Mops
Fournisseur: FG CLEAN WIPES TE
New Product
Certificats
À propos de cet article
Optimized cleaning performance and efficiency. Mops are available in non sterile or validated sterile formats.
Removes 99.9% of particles down to 0.1 µm in size
- 28 m²/ 305 sq. ft. coverage per mop, 47% more than leading competitors
- Pocket mop design facilitates hands-free operation and minimizes the spread of dirty cleaning solutions
- High absorption and coverage to maximize worker efficiency
- Consistent and even laydown of cleaning solutions
- Certificate of sterility included with sterile mops
- 3-year shelf life
Microfiber cleaning power coupled with the hands-free mop drop system provides consistent cleanliness and minimizes the spread of dirty cleaning solutions while improving productivity. The exacting sterilization process offers absolute confidence for use with sterile mops.
Packaging:
Mops are packaged 5 per bag, 20 bags per case.
Frequently Bought Together