ContactUsLinkComponent
Saturix Stainless Steel Cart with Charging Bucket
Catalog # 77871-058
Supplier:  FG CLEAN WIPES TE
New Product
Saturix Stainless Steel Cart with Charging Bucket
Catalog # 77871-058
Supplier:  FG CLEAN WIPES TE
Supplier Number:  HC102
New Product
Specifications
  • Dimensions:
    24L×25W×40"H
  • Material:
    Stainless steel
Specifications
About this item
Risk reduction for better Annex 1 compliance. A complete, patented system for critical cleaning engineered to work flawlessly together or alone to reduce risk and waste, promote employee health and safety, and improve operational efficiency.
  • 316L electropolished stainless steel
  • Recess-free cleanroom welds
  • Extreme durability with easy cleaning
  • Efficient batch preparation for up to 10 mops
  • Hands-free mop attachment and removal to minimize the spread of dirty cleaning solutions
  • Charging buckets with arched bottom for easy frame insertion
  • Modular design enables easy upgrade to premium options
  • Seamless integration with Saturix handles, frames, and mops
This entry-level cart offers superior performance features over traditional options, including a modular design, patented arched charging buckets, and the Hands-Free Mop Drop System.