Saturix Stainless Steel Cart with Charging Bucket
Catalog # 77871-058
Supplier: FG CLEAN WIPES TE
New Product
Specifications
- Dimensions:24L×25W×40"H
- Material:Stainless steel
Specifications
About this item
Risk reduction for better Annex 1 compliance. A complete, patented system for critical cleaning engineered to work flawlessly together or alone to reduce risk and waste, promote employee health and safety, and improve operational efficiency.
316L electropolished stainless steel
- Recess-free cleanroom welds
- Extreme durability with easy cleaning
- Efficient batch preparation for up to 10 mops
- Hands-free mop attachment and removal to minimize the spread of dirty cleaning solutions
- Charging buckets with arched bottom for easy frame insertion
- Modular design enables easy upgrade to premium options
- Seamless integration with Saturix handles, frames, and mops
This entry-level cart offers superior performance features over traditional options, including a modular design, patented arched charging buckets, and the Hands-Free Mop Drop System.