Saturix Sterile Isopropyl Alcohol
Supplier: FG CLEAN WIPES TE
New Product
Certificates
About this item
The dependability and performance of Saturix sterile alcohol spray is unmatched.
70% USP grade Isopropyl Alcohol
- 30% water for injection (WFI) quality
- Alcohol blend filtered to 0.1 µm
- Each bottle is double bagged
- Pyrogen-free
- Gamma irradiated and validated to a sterility level of 10⁻⁶
Fast-acting with extremely low chemical residue, this sterile IPA is manufactured in an ISO 6 cleanroom and packaged in an ISO 5 cleanroom, according to the highest GMP and ISO 9001 standards.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together