À propos de cet article
The ISO NOP007 has a tip diameter of just 7 µm and a length of 2 mm. The response is linear over a wide dynamic concentration range of NO.
Excellent selectivity to NO
Rapid response time
Highly sensitive
Response time: <3 seconds
Poise voltage: 865 mV
Linear response over a large dynamic concentration range of NO
Dry electrodes require no sleeves or filling solution
The design is based on a single carbon fiber coated with WPI's no-selective membrane. A detection limit of approximately 1 nM no makes this electrode ideal for use in tissues and microvessels.
The ISO NOP30 has a tip diameter of 30 microns and is available in two different tip lengths ISO NOP3020 has tip length of 2 mm, ISO-NO3005 has tip length of 0.5 mm). The response is linear over a wide dynamic concentration range of no. The design is based on a single carbon fiber coated with WPI's no-selective membrane. A detection limit of approximately 1 nM no makes these electrodes ideal for use in tissues and microvessels.
Requires cable (77663-980) sold separate.
Package of 3.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.