Glass Fiber Extraction Thimbles, Grade G400
New Product
These glass fiber thimble are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
Composed of glass fiber cellulose for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Glass fiber thimble offer remarkable resistance to high temperatures
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
They are well suited to high temperature applications and are free of binders with high purity.
Frequently Bought Together