ContactUsLinkComponent
ICT+ Isolator Cleaning Tool, Handle and Frame Kit
Catalog # 77871-012
Supplier:  FG CLEAN WIPES TE
New Product
undefined
ICT+ Isolator Cleaning Tool, Handle and Frame Kit
Catalog # 77871-012
Supplier:  FG CLEAN WIPES TE
Supplier Number:  IFHTA
New Product
Specifications
  • Description:
    Isolator cleaning tool
Specifications
About this item
The Isolator Cleaning Tool will help any compounding pharmacy be more compliant with USP and ANNEX 1 standards to ensure the clean and sterile preparation of drugs for the safety of patients and employees.
  • Touch-free pad application and removal; clean pads click on the frame, dirty pads click off the frame
  • Reduces cross contamination risk while increasing efficiency
  • Ergonomic, easy-to-use hybrid design
  • Asymmetrical frame maximizes coverage and effectively cleans tight spaces
  • Offset yoke on the frame extends reach, making it easy to clean high areas
  • 99.9% particulate residue removal; microfiber cleaning pads remove 99.9% of particles down to 0.1 micron
  • Mechanically removes disinfectant residues and other harmful particulates from the surface
Conventional isolator cleaning tools require extensive handling to attach and remove pads, which takes time and poses a significant risk for cross contamination. The ICT+ isolator cleaning tool, with patent-pending designs, not only features touch-free operation, but also provides an efficient system for used pad management with superior overall usability. The ICT+ isolator cleaning tool is a versatile tool for systematic and hygienic cleaning of isolators, biosafety cabinets, laminar flow hoods, and other aseptic environments.