Butyl Gloves, 20 mil
Supplier: PIP
New Product
Certificates
About this item
These 14" extended length 20 mil butyl rubber gloves are available with a smooth grip or rough grip finish.
Versatile seamless butyl glove
- Delivers superior resistance against aldehydes, ketones, esters, and concentrated mineral acids
Ideal for laboratory, aircraft maintenance, automotive, propellant manufacturing and munitions handling applications.
Specifications
- Catalog No:
- 77874-582
- 77874-590
- 77874-594
- 77874-588
- 77874-580
- 77874-592
- 77874-596
- 77874-610
- 77874-598
- 77874-608
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +