CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
Supplier:   Beckman Coulter
CytoFLEX Mosaic Detection Modules
Supplier:  Beckman Coulter
Certificates
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
14 Options Available Below
Product Details & Documents
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful CytoFLEX flow cytometry solution to date.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R-I (6 laser NUV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20163
  • CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R (5 laser NUV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20164
  • CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system V-B-Y-R (4 laser NUV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20165
  • CytoFLEX S and CytoFLEX Mosaic 63 spectral flow analyzer system B-R-V-Y (4 laser 561)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20166
  • CytoFLEX Mosaic 88 spectral detection module U-V-B-Y-R-I (6 laser CytoFLEX LX UV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19581
  • CytoFLEX Mosaic 88 spectral detection module U-V-B-Y-R (5 laser CytoFLEX LX UV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19582
  • CytoFLEX Mosaic 88 spectral detection module U-V-B-R (4 laser CytoFLEX LX UV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19583
  • CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R-I (6 laser CytoFLEX LX NUV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19584
  • CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R (5 laser CytoFLEX LX NUV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19585
  • CytoFLEX Mosaic 88 spectral detection module V-B-Y-R (4 laser CytoFLEX LX NUV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19586
  • CytoFLEX Mosaic computer and monitor
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D19589
  • CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R-I (6 laser UV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20160
  • CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R (5 laser UV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20161
  • CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-R (4 laser UV)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Minimum order quantity is 1
    Supplier Number: D20162