Human Recombinant SDF-1 A/CXCL12 (from E. coli cells)
New Product
Stromal cell-derived factor-1 alpha (SDF-1 α), also called CXCL12a, is one of two SDF-1 splice variants made by a wide variety of cells upon stimulation by inflammatory cytokines such as TNF, IL-1, and LPS. SDF-1α signals through the G protein-coupled receptor CXCR4 to recruit activated leukocytes. Human and mouse SDF-1 α share 99% sequence identity.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P48061
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL12|PBSF
- Protein/peptide name:SDF-1 A/CXCL12
- Purity:≥95%
- Molecular weight:8 kDa
- Sequence:KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together