Order Entry
Canada
ContactUsLinkComponent
Human Recombinant GROWTH HORMONE (from E. coli)
Human Recombinant GROWTH HORMONE (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant GROWTH HORMONE (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P01241
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Somatotropin
  • Protein/peptide name:
    GROWTH HORMONE
  • Purity:
    ≥95%
  • Molecular weight:
    22.3 kDa
  • Sequence:
    MFPTIPLSRL FDNAMLRAHR LHQLAFDTYQ EFEEAYIPKE QKYSFLQNPQ TSLCFSESIP TPSNREETQQ KSNLELLRIS LLLIQSWLEP VQFLRSVFAN SLVYGASDSN VYDLLKDLEE GIQTLMGRLE DGSPRTGQIF KQTYSKFDTN SHNDDALLKN YGLLYCFRKD MDKVETFLRI VQCRSVEGSC GF
  • Endotoxin level:
    Low
  • Formulation:
    20 mM sodium bicarbonate, pH 8.0
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together