Order Entry
Canada
ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike RBD IgA Titer Serologic Assay Kit
Human Anti-SARS-CoV-2 Spike RBD IgA Titer Serologic Assay Kit
Catalog # 77761-658
Supplier:  ReVacc Scientific
New Product
CAS Number:  
undefined
Human Anti-SARS-CoV-2 Spike RBD IgA Titer Serologic Assay Kit
Catalog # 77761-658
Supplier:  ReVacc Scientific
Supplier Number:  D71N96
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Conjugate ELISA:
    Unconjugated
  • Format:
    Pre-coated
  • Host:
    Goat
  • Primary antibody reactivity:
    Human
  • Target protein:
    Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) spike receptor-binding domain (RBD) subunit
  • Size:
    96 tests

Specifications

About this item

This kit is developed for serologic test for human IgA titer of Anti-SARS-CoV-2 trimeric Spike (Wuhan Strain) antibody in serum/plasma or purified human antibody (monoclonal and polyclonal) in vitro. Antibodies of IgM and IgA class are a sign of an active infection (primary infection and reactivation) and disappear during convalescence. In some cases they can persist for several months.

  • These kits were tested to bind to monoclonal antibodies in IgA format (Q74F36) or human sera

  • Recommendations - customers who intend to use this kit to generate quantitative curves are suggested to consider the monoclonal antibodies to generate the reference curve: Recombinant Human Anti-SARS or SARS-CoV-2 S RBD IgA Antibody (Q74F36)

Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) caused the pandemic of COVID-19. The receptor-binding domain (RBD) subunit of the spike protein is to bind to its receptor on target cell surface (ACE2). A rapid and effective assay kit detecting the levels of anti-SARS-CoV-2 Spike RBD in human serum can facilitate research on antibodies produced in response to SARS-CoV-2 infection or vaccines.

Assay principles:
This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti- Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) RBD subunit of Spike (Wuhan Strain) human IgA in serum. The kit consists of high-bind detachable 96-well plate pre-coated with Spike RBD subunit protein, positive control, negative control, an HRP-Anti-Human IgA secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (Four simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells four times by wash solution (0.3 ml/well)
2. Dilute the Secondary antibody HRP-Anti-Human IgA by wash solution and add to each well (0.1 ml/well). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml/well) and incubate at room temperature for 10 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD Value reflects the amount of antibody bound.

Caution:

For research use only.