Zero Waste Box™ Laboratory PPE and Plastic Recycling Boxes
Fournisseur: TERRACYCLE CANADA ULC CA
New Product
Certificats
À propos de cet article
This easy-to-use recycling program provides a complete waste collection, shipment, and recycling solution for disposable PPE (face masks, gloves, garments etc), protective eyewear (safety glasses), rigid lab plastics, flexible plastics, and disinfectant wipes.
Reduce landfill waste
- Support your company's sustainability goals
- Each collection box can be placed at convenient locations throughout the lab or facility
- When full, simply place the collection box in a designated shipping area
- Each box has a pre-paid UPS shipping label affixed
This program offers a convenient opportunity to recycle various types of waste found in most labs. Any item contaminated with hazardous, flammable, or infectious substances are not accepted in this box.
Each sturdy collection box is made of double-corrugated cardboard. Boxes are supplied with simple assembly instructions and a pre-affixed return shipping label.
Spécifications
- Catalog No:
- CA77867-916
- CA77867-912
- CA77867-914
- Environmentally Preferable:
- Waste Reducing, Sustainable Materials
- Waste Reducing, Sustainable Materials
- Waste Reducing, Sustainable Materials
- Environmental certification:
- FSC (Licence Code FSC-C018029), Bureau Veritas
- FSC (Licence Code FSC-C018029), Bureau Veritas
- FSC (Licence Code FSC-C018029), Bureau Veritas