Extended Length LfTS fit Pipette Tip Reload Systems, Non Sterile
77873-988
: VistaLab
New Product
- Volume:1 - 10 µl
- Length:50.04 mm
- Tip Color:Clear
- Packed:960 tips/reload
- Sterile:-
- Tip Style:Extended length
- For Use With:LfTS, LTS
Designed to provide a low-force tip solution for more comfortable pipetting; less force needed to acquire tips.
LfTS reload systems offer a convenient and economical way to refill empty tip racks, saving both time and money
- Reload systems dramatically reduce plastic waste by reusing empty racks
- Fine tip for improved accuracy and precision
- Polypropylene
These 10 μl extended length tips have a maximum volume of 20 μl and are compatible with 2, 10, and 20 μl pipettes. LfTS reload system tips, wafers, and racks are fully autoclavable.