5-Methyl-Cytidine-5-Triphosphate (5-Methyl-CTP)
Catalogue # 77868-090
Fournisseur: New England Biolabs (NEB)
New Product
Numéro CAS:
Spécifications
- Description:5-Methyl-Cytidine-5-Triphosphate (5-Methyl-CTP), 100 mm concentration
- Volume:0.1 ml
Spécifications
À propos de cet article
mRNA has emerged as a potent modality for therapeutics.
Supplied as sodium salts, pH 7.0
- Modified NTPs are commonly used for reduction of immunogenicity for in vitro transcription RNA
- No detectable RNase, DNase, phosphatase or protease contaminating activities
- ≥95% purity, as determined by HPLC
-
Optimal activity and stability for up to 24 months
Concentration: 100 mM
Incorporation of chemically modified ribonucleotides improves the efficacy of these therapies by increasing stability, reducing immunogenicity, and enhancing translatability of mRNAs.