Corepoint Scientific™ Temperature Test Chambers 72 Cu. Ft., Horizon Scientific
Fournisseur: Horizon Scientific
New Product
Certificats
À propos de cet article
Transform your product testing with Corpoint® Scientific Temperature Test Chambers.
- Programmable PID controller
- Variable speed compressor
- Broad temperature range
- USB data port
- Natural hydrocarbon refrigerants
Ensure the quality and durability of your products with Corepoint® Scientific Temperature Test Chambers. These chambers provide a controlled temperature environment to deliver superior product testing backed by FDA and ICH standards. Improve product quality, test packaging, increase shelf life, and enhance safety with these chambers. With natural refrigerants and smart controllers, these units feature some of the most advanced technology in the industry.
Certifications: ETL/cETL Listed
Spécifications
- Catalog No:
- 77873-420
- 77873-416
- Int. W×D×H:
- 1695×594×1511 mm (66.75×23.375×59.5")
- 1695×594×1511 mm (66.75×23.375×59.5")
- Ext. W×D×H:
- 2057×921×2070 mm (81×36.25×81.5")
- 2057×921×2070 mm (81×36.25×81.5")