Anti-VEEV Human Recombinant Monoclonal Antibody [clone: RV24]
Fournisseur: ReVacc Scientific
New Product
Certificats
À propos de cet article
This antibody has been tested to bind to Venezuelan Equine Encephalitis Virus (VEEV) envelope (E) glycoprotein (E3, E2 and E1 with 6K) (Cat. No 77761-732).
Type: Primary
Antigen: VEEV
Clonality: Recombinant Monoclonal
Clone: RV24
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 kappa
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77761-688
- 77761-690
- Antigen symbol:
- VEEV
- VEEV
- Antigen name:
- Venezuelan Equine Encephalitis Virus
- Venezuelan Equine Encephalitis Virus
- Antigen synonyms:
- Envelope (E) glycoprotein (E3, E2 and E1 with 6K)
- Envelope (E) glycoprotein (E3, E2 and E1 with 6K)
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV24
- RV24
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Molecular weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by Refractometry
- 1 mg/ml by Refractometry
- Storage buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping temperature:
- Ice
- Ice