Compact Dot Matrix Printer
Supplier:   Sartorius
Compact Dot Matrix Printer
Supplier:  Sartorius
Certificates
2 Options Available Below
Product Details & Documents
Dot matrix printer for Sartorius balances.
  • Compact and robust design
  • High printing speed
Acquisition and documentation of current measurement data.
Specifications for Product Family
Specifications
  • Catalog No:
  • CA77870-884
  • CA77870-886
  • Width:
  • 114.6 mm
  • 120 mm
  • Depth:
  • 188.2 mm
  • 315 mm
  • Height:
  • 88.6 mm
  • 70 mm
  • Weight:
  • Net: 1.5 kg; Gross: 2 kg
  • Net: 1 kg; Gross: 1.06 kg
Specifications
Product Family Options
Printer
Product Information
  • Désignation
    Availability
    Price
  • Compact dot matrix printer
    Log in for availability
    Log in for availability
    Each
    Each - $902.00
    $902.00
    Each
       $902.00
    Supplier Number: YDP50
    Specifications:
    More
    Specifications
    Cat. No.
    CA77870-884
    Width
    114.6 mm
    Depth
    188.2 mm
    Height
    88.6 mm
    Weight
    Net: 1.5 kg; Gross: 2 kg
Printer paper
Product Information
  • Désignation
    Availability
    Price
  • Printer paper, YDP50, 57×50 mm (10×)
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $53.35
    $53.35
    Pack of 10
       $53.35
    Supplier Number: YDP50-PAPER
    Specifications:
    More
    Specifications
    Cat. No.
    CA77870-886
    Width
    120 mm
    Depth
    315 mm
    Height
    70 mm
    Weight
    Net: 1 kg; Gross: 1.06 kg