Hanna pH Buffer Sachets | Avantor
Hanna pH Buffer Sachets
Supplier:   Hanna
Hanna pH Buffer Sachets
Supplier:  Hanna
Certificates
Hanna pH Buffer Sachets
Hanna pH Buffer Sachets
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This buffer value is widely used in water purification plants, in the food industry, and wherever the pH is expected to be slightly acidic.
  • Basic buffers that suit most users
  • Certificate of Analysis (COA) on request
  • Accuracy of ±0.01 pH at 25 °C
  • Traceable to NIST standard
Each sealed sachet holds just the right amount of solution making every sachet as fresh as the day it was packaged. Perfect for use with testers and portable meters. Multiple resolutions for better accuracy.
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    No. of Tests
    Availability
    Price
  • pH 4 buffer (sachet)
    20 ml
    25 tests
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $42.50
    $42.50
    Log in pour la disponibilité
    Each
    Each - $0.00
    $0.00
    Case of 25
       $42.50
    Each
       $0.00
    Supplier Number: HI70004P
  • pH 7 buffer (sachet)
    20 ml
    25 tests
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $42.50
    $42.50
    Log in pour la disponibilité
    Each
    Each - $0.00
    $0.00
    Case of 25
       $42.50
    Each
       $0.00
    Supplier Number: HI70007P