About this item
FG Cleanroom Saturix 3400SE wipes are 100% polyester with sealed edges for minimal particle release. Designed for critical environments, they provide exceptional cleanliness, durability, and absorbency, ensuring effective contamination control in cleanrooms, labs, and precision manufacturing applications.
100% polyester knit
- Sealed edges
- Heavyweight construction, 145 gsm
- Double bagged in heavy mil, poly cleanroom packaging
- Processed and packed in an ISO 4 cleanroom
- High durability and chemical resistance
- Economy and performance
- Low NVRS, fibers, ions, and extractables
The FG Cleanwipes Saturix Poly Wipes are high-performance cleanroom wipes crafted from 100% polyester, designed for superior cleanliness, strength, and contamination control in critical environments. These wipes feature low-linting properties, excellent durability, and exceptional absorbency, making them ideal for wiping surfaces, absorbing spills, and removing contaminants without shedding fibers. Their advanced design ensures minimal particulate release, supporting cleanroom compliance and product integrity. The FG Saturix Poly Wipes provide a reliable, cost-effective solution for industries that demand the highest standards of cleanliness, durability, and operational efficiency.
Target industries include:
Pharmaceutical and Biotech: ideal for sterile manufacturing, surface wiping, and contamination control.
Electronics and Semiconductor: ensures pristine, particulate-free surfaces during assembly and precision cleaning.
Medical Device Manufacturing: supports stringent cleanliness standards for cleanrooms and production areas.
Aerospace and Automotive: effective for maintaining contaminant-free environments in sensitive manufacturing processes.