Proteometer-UFT MD Unit | Avantor
Proteometer-UFT MD Unit
Proteometer-UFT MD unit
Catalog # 77775-502
Supplier:   NOVILYTIC, LLC
Fournisseur #:  RCT-PUF-001
Proteometer-UFT MD Unit
Catalog # 77775-502
Supplier:  NOVILYTIC, LLC
Proteometer-UFT MD Unit
Proteometer-UFT MD Unit
Proteometer-UFT MD unit
Price
$970.75
Log in pour la disponibilité
Each
Product Details & Documents
Proteometer-UFT MD unit sold individually.
  • Proteometer-UFT MD unit replaces the LC column in the analytical system
Intended for use with the Proteometer-UFT kit.
 : Reorder individually as needed.
 : Supplied in a custom box with instructions for use. Follow instructions in "Instructions for Use" for proper storage.
Spécifications
  • Désignation:
    Proteometer-UFT MD unit