Precision II Tubing Rollstock offers high-quality, customizable packaging solutions designed for industries requiring durability and precision. Engineered for secure sealing and reliable performance, this rollstock is ideal for applications in healthcare, food, and industrial sectors. It ensures efficient production while maintaining product integrity and compliance with industry standards.
Manufactured in an ISO Class 4 Cleanroom
- Manufactured from ultrapure clean low density polyethylene resin
- Customizable sizing: Flexible rollstock allows users to create custom packaging solutions tailored to specific needs
- Durable material: Provides excellent strength and durability for secure and reliable containment
- Convenient and versatile
Precision II Cleanroom Tubing Rollstock offers high-quality, contamination-controlled packaging solutions designed for cleanroom and critical environments. Precision II Tubing Rollstock is perfect for packaging sterilized instruments, sensitive electronic components in controlled environments and ideal for industries requiring contamination-free packaging, including pharmaceuticals, medical devices, and electronics. It supports compliance with stringent cleanliness and quality standards, ensuring product integrity.
Certified to KSC-C-123J; Cleanroom class 100.