VWR® Dissolved Oxygen Electrodes, Avantor
Fournisseur: SCHULER SCIENTIFIC
New Product
Certificats
À propos de cet article
The VWR® dissolved oxygen electrodes allow for the measurement of oxygen within a solution.
Optical or polarographic sensor technology options
- Built-in temperature probe
- For use with VWR® dissolved oxygen meters
For application specific recommendations, please reference "VWR® electrode selection guide" located in the documents section.
Frequently Bought Together