ContactUsLinkComponent
VWR® 100 Multi-Parameter Pocket Tester (pH/ORP/Cond./TDS/Sal./Res.), Avantor
  77619-092
New Product
undefined
VWR® 100 Multi-Parameter Pocket Tester (pH/ORP/Cond./TDS/Sal./Res.), Avantor
  77619-092
New Product
  • Model:
    E-M100
  • Description:
    Multi-parameter pocket tester, pH/conductivity/ORP sensor
  • Parameters:
    pH/ORP/conductivity/TDS/salinity/resistivity
The VWR 100-series multi-parameter pocket tester is a versatile system that provides pH, conductivity, and ORP measurements with a single tester.
  • High resolution LCD display
  • Auto and manual HOLD function: Freezes readings on the display for easy and accurate recording
  • IP67 waterproof
  • Automatic temperature compensation capabilities
  • Backlight adjustment and automatic shutdown: Extends battery service life
  • Replaceable electrode
The easy to use VWR-M100 multi-parameter pocket tester provide the versatility of multiple measurement parameters in a single tester. The rugged carrying case allows you to easily transport the pocket tester, buffers and accessories to any location. Easily switch out the electrode to help prolong the life of the pocket tester.
 :
Tester includes: Carrying case, wrist strap, pH buffers (4.00 and 10.01), conductivity standards (1413 µS/cm and 12.85 mS/cm), ORP calibration solution (222 mV), calibration bottles, AAA batteries (4).