VWR® 950 Benchtop pH Meter
Supplier:   VWR International
VWR® 950 Benchtop pH Meter
Supplier:  VWR International
Certificates
VWR® 950 Benchtop pH Meter
VWR® 950 Benchtop pH Meter
9 Options Available Below
Product Details & Documents
The VWR-PH950 meter provide accurate and reliable measurements of pH, mV, ORP and temperature.
  • High resolution color touch screen (7.0")
  • 1 to 6 point calibration with standard recognition
  • Selectable pH buffer groups, including USA, NIST, DIN, GB, MERK, JIS
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 1000 sets of measurement results (GLP-compliant)
  • Automatic electrode diagnosis with pH slope and offset display
The easy to use 77619-222 benchtop meters provides accurate measurements in the field and the laboratory. The intelligent operation system provides features including user management, calibration management, electrode management, method management, and many other advanced capabilities. Connect the meter easilty to the VWR Dot Matrix Printer (10799-132) to print measuremetn reports.
For application specific recommendations, please reference 'VWR® electrode selection guide' located in the related information section.
 : IP 54 protection.
 : Three-year meter warranty.
 : All meters include: Free standing electrode holder, silicone rubber In-use cover, and touch screen pen.
Product Family Options
No electrode
Informations sur le produit
  • Désignation
    Availability
    Price
  • VWR-PH950 meter, no electrode
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,069.02
    $2,069.02
    Prix promotionnel pour les nouveaux clients
    Each
       $2,069.02
Electrode included
Informations sur le produit
  • Désignation
    Availability
    Price
  • VWR-PH950 meter with PH31 3-in-1 composite general purpose electrode and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,303.24
    $2,303.24
    Prix promotionnel pour les nouveaux clients
    Each
       $2,303.24
  • VWR-PH950 meter with PH49 strong alkali resistance electrode, TEMP01 ATC probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,558.42
    $2,558.42
    Prix promotionnel pour les nouveaux clients
    Each
       $2,558.42
  • VWR-PH950 meter with PH32 premium TRIS-compatible electrode, TEMP01 ATC probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,381.34
    $2,381.34
    Prix promotionnel pour les nouveaux clients
    Each
       $2,381.34
  • VWR-PH950 meter with PH44 low maintenance electrode, TEMP01 ATC probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,558.42
    $2,558.42
    Prix promotionnel pour les nouveaux clients
    Each
       $2,558.42
  • VWR-PH950 meter with PH47 ultrapure water electrode, TEMP01 ATC probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,782.70
    $2,782.70
    Prix promotionnel pour les nouveaux clients
    Each
       $2,782.70
  • VWR-PH950 meter with PH38 blade spear electrode, TEMP01 ATC Probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,808.20
    $2,808.20
    Prix promotionnel pour les nouveaux clients
    Each
       $2,808.20
  • VWR-PH950 meter with PH36 slender electrode, TEMP01 ATC probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,625.86
    $2,625.86
    Prix promotionnel pour les nouveaux clients
    Each
       $2,625.86
  • VWR-PH950 meter with PH40 cleanable electrode, TEMP01 ATC probe, and pH buffers (4.01, 7.00, 10.01) 50 ml bottles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,558.42
    $2,558.42
    Prix promotionnel pour les nouveaux clients
    Each
       $2,558.42
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Replacement pen for use with touchscreen
    Replacement pen for use with touchscreen
    77618-916
    VWR® 900-, 950-series meters
    Log in pour la disponibilité
    Each - $19.74
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Magnetic stirrer
    Magnetic stirrer
    77618-914
    All series
    Log in pour la disponibilité
    Each - $394.87
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Replacement pH buffers (4.01, 7.00, 10.01 pH), 50 ml each
    Replacement pH buffers (4.01, 7.00, 10.01 pH), 50 ml each
    77618-908
    All series
    Log in pour la disponibilité
    Pack of 3 (50ml) - $30.71
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Replacement conductivity standard (1413 µS/cm), 50 ml
    Replacement conductivity standard (1413 µS/cm), 50 ml
    77618-826
    All series
    Log in pour la disponibilité
    Each(1/Itm/Bx) - $17.54
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Replacement screen cover
    Replacement screen cover
    77618-812
    VWR® 950-series meters
    Log in pour la disponibilité
    Each - $26.33
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Replacement electrode stand, free standing
    Replacement electrode stand, free standing
    77618-912
    All series
    Log in pour la disponibilité
    Each - $65.82