VWR® pH Electrodes
À propos de cet article
The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.
Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements
- BNC connector (BNC(Q9) or S7)
- Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
- Gel or liquid electrolyte fill solution
- Includes unique serial number
- Easily connect to VWR(R) pH meters
For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.
20 Options Available Below
Product Details & Documents
The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.
Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements
- BNC connector (BNC(Q9) or S7)
- Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
- Gel or liquid electrolyte fill solution
- Includes unique serial number
- Easily connect to VWR(R) pH meters
For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.
Product Family Options
Informations sur le produit
- DésignationTypeMatièreConnecteurFill SolutionPlage de températurepH RangeAvailabilityPrice
- 3-in-1 pH composite electrode (built-in ATC)PolycarbonateBNC(Q9) and 4-pin aviation3 M KCl5...60 °C0 - 14Log in pour la disponibilitéEach$254.50
- pH composite electrode (Long)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...80 °C0 - 14Log in pour la disponibilitéEach$505.43
- pH composite electrode (Blade spear)POMBNC(Q9)Unfillable5...60 °C0 - 10Log in pour la disponibilitéEach$173.74
- pH composite electrode (Low maintenance)PolycarbonateBNC(Q9)Unfillable0...80 °C0 - 14Log in pour la disponibilitéEach$515.40
- pH composite electrode (High Temp. resistance)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...100 °C0 - 14Log in pour la disponibilitéEach$515.40
- pH composite electrode (RoHS)Lead-free glassS7 [S7-BNC(Q9) cable]3 M KCl0...80 °C0 - 14Log in pour la disponibilitéEach$515.40
- pH composite electrode (Ultrapure water)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...80 °C0 - 11Log in pour la disponibilitéEach$751.62
- pH composite electrode (HF acid resistance)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...100 °C0 - 14Log in pour la disponibilitéEach$515.40
- pH composite electrode (Alkali resistance)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...80 °C0 - 14Log in pour la disponibilitéEach$515.40
- pH composite electrode (HF acid titrate)PolypropyleneS7 [S7-BNC(Q9) cable]3 M KCl0...60 °C2 - 11Log in pour la disponibilitéEach$505.43
- pH composite electrode (TRIS compatible)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...80 °C0 - 14Log in pour la disponibilitéEach$359.60
- pH composite electrodePolycarbonateBNC(Q9)Unfillable5...60 °C0 - 14Log in pour la disponibilitéEach$123.73
- pH composite electrodePolycarbonateBNC(Q9)3 M KCl5...60 °C0 - 14Log in pour la disponibilitéEach$123.73
- pH composite electrode (Slender)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...60 °C0 - 14Log in pour la disponibilitéEach$586.44
- pH composite electrode (Flat)GlassS7 [S7-BNC(Q9) cable]3 M KCl0...60 °C1 - 12Log in pour la disponibilitéEach$586.44
- pH composite electrode (Blade spear)POM and 316 Stainless SteelBNC(Q9)3 M KCl0...60 °C0 - 14Log in pour la disponibilitéEach$763.42
- pH composite electrode (Low conductivity)EpoxyS7 [S7-BNC(Q9) cable]Unfillable0...80 °C0 - 11Log in pour la disponibilitéEach$373.81
- pH composite electrode (Cleanable)EpoxyBNC(Q9)3 M KCl0...80 °C0 - 14Log in pour la disponibilitéEach$505.43
- pH composite electrode (Flat)PolycarbonateBNC(Q9)3 M KCl5...60 °C0 - 14Log in pour la disponibilitéEach$243.86
Accessories
Informations sur le produit
- Catalog #MaterialFor Use WithAvailabilityPrice
- VWR® ATC probeVWR® ATC probe77618-918316 stainless steelVWR® 500-, 550-, 700-, 750-, 900, 950-series metersLog in pour la disponibilitéEach - $163.22