Infrared Thermometer Guns, 12:1/932 ºF
Fournisseur: Sper Scientific
New Product
Certificats
À propos de cet article
Accurate IR at a reasonable price.
Backlit LCD
- Laser guide
- Min./max.
- Audible high/low alarms
- Memory
IR thermometers are used to safely measure the temperature in: Inaccessible, hot or hazardous objects in electrical, hazmat, HVAC, and other fields; moving machinery in factories, automobiles and elsewhere; electrical troubleshooting HVAC energy audits; avoiding contamination in the food, beverage and pharmaceutical industries.
Ordering information:
Certification is a NIST traceable certificate showing compliance with the published specifications for the product.
Spécifications
- Catalog No:
- 470357-590
- 470357-592
- Emissivity:
- Adjustable 0.1 to 1.0
- Adjustable 0.1 to 1.0