Logic® Class II, Type A2 Biosafety Cabinets
Logic® Class II, Type A2 Biosafety Cabinets
Supplier:   Labconco
Logic® Class II, Type A2 Biosafety Cabinets
Supplier:  Labconco
Certificates
Logic® Class II, Type A2 Biosafety Cabinets
Logic® Class II, Type A2 Biosafety Cabinets
4 Options Available Below
Product Details & Documents
REDISHIP Logic Class II, Type A2 Biosafety Cabinets with 10" sash, right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand, 115V, 60Hz
  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Logic OS with 5” line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
  • Telescoping base stand with shelf and leveling feet
  • Full, six-year warranty on all Class II biosafety cabinets
Logic Class II, Type A2 Biosafety Cabinets (BSCs) offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1-4 containment. These cabinets are ideal for applications involving biological hazards, genetic material, antineoplastic drugs and other hazardous airborne particulates. All models ensure the highest standard of safety and performance and are listed to NSF/ANSI 49. Designed for application versatility, Logic A2 BSCs either exhaust HEPA-filtered air into the laboratory or can be connected to a laboratory exhaust system with applications involving chemical vapors or odors.
Logic BSCs maximize safety, performance, and comfort for users. At the core is Constant Airflow Profile™ (CAP) technology, which keeps users and their samples protected at all times. Now paired with BalanceFlow™ plenum technology, Logic cabinets use up to 30% less energy and are up to 5 decibels quieter. Every Logic is powered by Logic OS – an advanced software platform with touchscreen interface located right at eye-level within the cabinet.
Certifications: NSF/ANSI Standard 49 listed
ISO Class 5 (Grade A) per ISO 14644-1
UL Standard 61010-1
CAN/CSA 22.2 No. 61010-1
Product Family Options
Informations sur le produit
  • Désignation
    Electrical
    Profondeur
    Hauteur
    Width
    External Dimensions
    Sash Opening
    Shipping Weight
    Availability
    Price
  • 3' REDISHIP Logic Class II, Type A2 Biosafety Cabinets with right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand
    115V, 60Hz, 12A
    81.3 cm (32")
    157.2 cm (61.9")
    107.7 cm (42.4")
    107.7×81.3×157.2 cm (42.4×32×61.9")
    10"
    697 lbs
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $21,570.00
    $21,570.00
    Each
       $21,570.00
    Supplier Number: 302319102
  • 4' REDISHIP Logic Class II, Type A2 Biosafety Cabinets with right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand
    115V, 60Hz, 12A
    81.3 cm (32")
    157.2 cm (61.9")
    138.2 cm (54.4")
    138.2×81.3×157.2 cm (54.4×32×61.9")
    10"
    808 lbs
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $24,310.00
    $24,310.00
    Each
       $24,310.00
    Supplier Number: 302419102
  • 5' REDISHIP Logic Class II, Type A2 Biosafety Cabinets with right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand
    115V, 60Hz, 16A
    81.3 cm (32")
    157.2 cm (61.9")
    168.7 cm (66.4")
    168.7×81.3×157.2 cm (66.4×32×61.9")
    10"
    953 lbs
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $27,100.00
    $27,100.00
    Each
       $27,100.00
    Supplier Number: 302519102
  • 6' REDISHIP Logic Class II, Type A2 Biosafety Cabinets with right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand
    115V, 60Hz, 16A
    81.3 cm (32")
    157.2 cm (61.9")
    199.1 cm (78.4")
    199.1×81.3×157.2 cm (78.4×32×61.9")
    10"
    1091 lbs
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29,960.00
    $29,960.00
    Each
       $29,960.00
    Supplier Number: 302619102