Platinum® Pro Barcoding Solutions, Quantum-Si
Reagents for protein barcoding. Generates 8 libraries for sequencing, 8
Catalog # 77778-314
Supplier:   QUANTUM-SI
 :  910-00047-01
Platinum® Pro Barcoding Solutions, Quantum-Si
Catalog # 77778-314
Supplier:  QUANTUM-SI
CAS Number:  
Platinum® Pro Barcoding Solutions, Quantum-Si
Platinum® Pro Barcoding Solutions, Quantum-Si
Reagents for protein barcoding. Generates 8 libraries for sequencing, 8
CAS Number:  
Price
$5,437.37
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Protein Barcoding with Next-Gen Protein Sequencing™. Next-Generation Protein Sequencing™ (NGPS™) offers single-molecule sequencing of multiplexed protein barcodes with quantitative results.
  • Drug delivery: Elevate and optimize drug delivery vehicles like LNPs or AAVs with protein barcodes
  • Protein engineering: Efficiently analyze protein characteristics in multiplex with protein barcodes
Advance drug delivery and protein analysis with precision barcoding. Characterize multiplexed drug delivery and translation efficiency of mRNA therapeutics with lipid nanoparticles (LNPs). Multiplexed analysis of target therapeutics and drug delivery systems accelerates screening cycles, reduces resources of in vivo and in vitro models, and saves time.
How it Works: Protein barcoding provides users with a robust method to screen multiple protein variants in a single pooled reaction. Individual proteins are tagged with short, unique peptide barcodes that identify each variant. With the barcoding kit, screened proteins are prepared and barcodes are released for sequencing on Platinum®.
Spécifications
Spécifications
  • Désignation:
    Reagents for protein barcoding. Generates 8 libraries for sequencing
  • No. of Reactions:
    8