Order Entry
Canada
ContactUsLinkComponent
Platinum® Pro Instrument, Quantum-Si
Platinum® Pro Instrument, Quantum-Si
Catalog # 77778-306
Supplier:  QUANTUM-SI
New Product
undefined
Platinum® Pro Instrument, Quantum-Si
Catalog # 77778-306
Supplier:  QUANTUM-SI
Supplier Number:  910-10033-00
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Platinum Pro Instrument for next-generation protein sequencing™
  • Size:
    305×660×356 mm (12×26×14")
  • Weight:
    20.5 kg (45 lbs.)

Specifications

About this item

Platinum Pro Instrument for next-generation protein sequencing™.

  • Accessible benchtop instrument enables protein sequencing with limited laboratory space

  • Intuitive touch screen interface unlocks advanced protein analysis for novice and experienced users
  • Scalable solution delivers sample insights with less than three hours hands on time and regular innovative enhancements
  • Versatile with cloud analysis and storage or local informatics option
  • Insight generating solution for divers protein applications with single-molecule amino acid resolution including: Protein identification, Protein variant and post translational modification analysis, antibody sequencing, and protein screening with barcoding

Platinum Pro makes next-generation protein sequencing accessible today and for the future, packed with capability across diverse applications. Platinum Pro opens access to single molecule insights for every lab, everywhere.

Certifications:

Conforms to ICES-003:2020 Issue 7, UL STD(s) 61010-1 & 61010-2-081, and is certified to CSA STD C22.2# 61010-1-12 & 61010-2-081. 
Conforms with low voltage directives 2014/35/EC; EMC Directive 2014/35/EU; ROHS Directive 2011/65/EU; WEEE Directive 2012/19/EU.    
Conformité Européenne (CE) Assessment meets requirements for all relevant EU and UK directives. Meets Waste Electrical and Electronic Equipment Directive (WEEE).

Ordering information:

Includes 12 months manufacturer's warranty including Platinum Analysis Software and cloud data storage.