Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant UCP3 (from E. coli) | Avantor

Human Recombinant UCP3 (from E. coli)

1 mg
Catalog # ABCA_AB79512-1MG
New Product
Supplier:   Abcam
Fournisseur #:  AB79512-1MG
Human Recombinant UCP3 (from E. coli)
Catalog # ABCA_AB79512-1MG
Supplier:  Abcam
New Product
CAS Number:  
Human Recombinant UCP3 (from E. coli)
Human Recombinant UCP3 (from E. coli)

1 mg

CAS Number:  
Price
$1,660.00
Log in pour la disponibilité
Each (1mg)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Recombinant Human UCP3 protein is a Human Fragment protein, in the 181 to 214 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, ELISA, WB.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Taille:
    1 mg
  • Form:
    Lyophilized
  • Protein/peptide name:
    UCP3
  • Protein synonyms:
    SLC25A9|UCP3|Putative mitochondrial transporter UCP3|Solute carrier family 25 member 9|Uncoupling protein-3|UCP 3
  • Purity:
    >90%
  • Concentration:
    1 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCA_AB79512-1MG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    Constituents: 2.68% PBS
  • Conjugation:
    Unconjugated
  • Tested applications:
    ELISA, SDS-PAGE, WB
  • Species:
    Human
  • Animal-free:
    N

Spécifications