Human Recombinant ARL11 (from E. coli)
About this item
Recombinant Human ARL11 protein is a Human Full Length protein, in the 1 to 196 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recombinant Human ARL11 protein is a Human Full Length protein, in the 1 to 196 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ABCA_AB109953-100U
- ABCA_AB109953-500U
- Protein/peptide name:
- ARL11
- ARL11
- Protein synonyms:
- ARLTS1|ADP-ribosylation factor-like protein 11|ADP-ribosylation factor-like tumor suppressor protein 1
- ARLTS1|ADP-ribosylation factor-like protein 11|ADP-ribosylation factor-like tumor suppressor protein 1
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Animal-free:
- N
- N
- Biological activity:
- Standard
- Standard
- Purity:
- >90%
- >90%
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Form:
- Liquid
- Liquid
- Formulation:
- pH: 8, Constituents: 30% Glycerol (glycerin, glycerine), 0.58% Sodium chloride, 0.32% Tris HCl, 0.08% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0.03% EDTA
- pH: 8, Constituents: 30% Glycerol (glycerin, glycerine), 0.58% Sodium chloride, 0.32% Tris HCl, 0.08% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0.03% EDTA
- Storage conditions:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Specifications
Product Family Options
Product Information
- TailleAvailabilityPrice
- Supplier Number: AB109953-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.ABCA_AB109953-100UProtein/peptide nameARL11Protein synonymsARLTS1|ADP-ribosylation factor-like protein 11|ADP-ribosylation factor-like tumor suppressor protein 1Protein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>90%Concentration0.5 mg/mlFormLiquidFormulationpH: 8, Constituents: 30% Glycerol (glycerin, glycerine), 0.58% Sodium chloride, 0.32% Tris HCl, 0.08% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0.03% EDTAStorage conditionsStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGENew Product - Supplier Number: AB109953-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.ABCA_AB109953-500UProtein/peptide nameARL11Protein synonymsARLTS1|ADP-ribosylation factor-like protein 11|ADP-ribosylation factor-like tumor suppressor protein 1Protein/peptide typeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedAnimal-freeNBiological activityStandardPurity>90%Concentration0.5 mg/mlFormLiquidFormulationpH: 8, Constituents: 30% Glycerol (glycerin, glycerine), 0.58% Sodium chloride, 0.32% Tris HCl, 0.08% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0.03% EDTAStorage conditionsStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGENew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



