Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant FGF5 (from E. coli)

Human Recombinant FGF5 (from E. coli)

New Product
Supplier:   Abcam
Human Recombinant FGF5 (from E. coli)
Supplier:  Abcam
New Product
Certificates
Human Recombinant FGF5 (from E. coli)

5 Options Available Below

  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Recombinant human FGF5 protein (Active) is a Human Full Length protein, in the 18 to 268 aa range, expressed in Escherichia coli, with ≥95% purity, ≤1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Spécifications

Spécifications

  • Catalog No:
  • ABCA_AB219125-500U
  • ABCA_AB219125-100U
  • ABCA_AB219125-10UG
  • ABCA_AB219125-50UG
  • ABCA_AB219125-1MG
  • Protein/peptide name:
  • FGF5
  • FGF5
  • FGF5
  • FGF5
  • FGF5
  • Protein synonyms:
  • Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
  • Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
  • Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
  • Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
  • Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin content:
  • <1 EU/µg
  • <1 EU/µg
  • <1 EU/µg
  • <1 EU/µg
  • <1 EU/µg
  • Biological activity:
  • Bioactive
  • Bioactive
  • Bioactive
  • Bioactive
  • Bioactive
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
  • Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
  • Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
  • Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
  • Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
  • Storage conditions:
  • Store at −20 °C
  • Store at −20 °C
  • Store at −20 °C
  • Store at −20 °C
  • Store at −20 °C
  • Shipping temperature:
  • 20
  • 20
  • 20
  • 20
  • 20
  • Tested applications:
  • FuncS, SDS-PAGE
  • FuncS, SDS-PAGE
  • FuncS, SDS-PAGE
  • FuncS, SDS-PAGE
  • FuncS, SDS-PAGE

Spécifications

Product Family Options

Informations sur le produit

  • Taille
    Availability
    Price
  • 500 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500µG)
    Each (500µG) - $2,980.00
    $2,980.00
    Each (500µG)
       $2,980.00
    Minimum order quantity is 1

    Supplier Number: AB219125-500UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB219125-500U
    Protein/peptide name
    FGF5
    Protein synonyms
    Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <1 EU/µg
    Biological activity
    Bioactive
    Purity
    ≥95%
    Form
    Lyophilized
    Formulation
    Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
    Storage conditions
    Store at −20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
    New Product
  • 100 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100µG)
    Each (100µG) - $944.00
    $944.00
    Each (100µG)
       $944.00
    Minimum order quantity is 1

    Supplier Number: AB219125-100UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB219125-100U
    Protein/peptide name
    FGF5
    Protein synonyms
    Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <1 EU/µg
    Biological activity
    Bioactive
    Purity
    ≥95%
    Form
    Lyophilized
    Formulation
    Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
    Storage conditions
    Store at −20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
    New Product
  • 10 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10µG)
    Each (10µG) - $512.00
    $512.00
    Each (10µG)
       $512.00
    Minimum order quantity is 1

    Supplier Number: AB219125-10UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB219125-10UG
    Protein/peptide name
    FGF5
    Protein synonyms
    Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <1 EU/µg
    Biological activity
    Bioactive
    Purity
    ≥95%
    Form
    Lyophilized
    Formulation
    Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
    Storage conditions
    Store at −20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
    New Product
  • 50 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (50µG)
    Each (50µG) - $656.00
    $656.00
    Each (50µG)
       $656.00
    Minimum order quantity is 1

    Supplier Number: AB219125-50UG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB219125-50UG
    Protein/peptide name
    FGF5
    Protein synonyms
    Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <1 EU/µg
    Biological activity
    Bioactive
    Purity
    ≥95%
    Form
    Lyophilized
    Formulation
    Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
    Storage conditions
    Store at −20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
    New Product
  • 1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1mg)
    Each (1mg) - $4,330.00
    $4,330.00
    Each (1mg)
       $4,330.00
    Minimum order quantity is 1

    Supplier Number: AB219125-1MG
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB219125-1MG
    Protein/peptide name
    FGF5
    Protein synonyms
    Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Animal-free
    N
    Endotoxin content
    <1 EU/µg
    Biological activity
    Bioactive
    Purity
    ≥95%
    Form
    Lyophilized
    Formulation
    Constituents: 0.58% Sodium chloride, 0.16% Sodium phosphate
    Storage conditions
    Store at −20 °C
    Shipping temperature
    20
    Tested applications
    FuncS, SDS-PAGE
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.