Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant EAF1 (from E. coli) | Avantor

Human Recombinant EAF1 (from E. coli)

50 µg
Catalog # ABCA_AB177621-50UG
New Product
Supplier:   Abcam
Fournisseur #:  AB177621-50UG
Human Recombinant EAF1 (from E. coli)
Catalog # ABCA_AB177621-50UG
Supplier:  Abcam
New Product
CAS Number:  
Human Recombinant EAF1 (from E. coli)
Human Recombinant EAF1 (from E. coli)

50 µg

CAS Number:  
Price
$923.00
Log in pour la disponibilité
Each (50µG)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Recombinant Human EAF1 protein is a Human Fragment protein, in the 182 to 268 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Taille:
    50 µg
  • Form:
    Liquid
  • Protein/peptide name:
    EAF1
  • Protein synonyms:
    ELL-associated factor 1|EAF1
  • Purity:
    >95%
  • Concentration:
    0.5 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCA_AB177621-50UG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0.88% Sodium chloride, 0.32% Tris HCl
  • Conjugation:
    Unconjugated
  • Tested applications:
    MS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.