Ham's F12/DME High Glucose 1:1 Mixture
Fournisseur: FUJIFILM IRVINE SCIENTIFIC INC.
New Product
Certificats
À propos de cet article
Ham’s F12/DME is a 1:1 mixture of Ham’s F12 and DMEM. This extremely rich, complex medium is commonly selected as the basal medium for the cultivation and study of challenging cell lines or primitive cells, such as neural progenitors.
Animal component-free
Chemically defined GMP manufacture
Spécifications
- Calcium:Yes
- Magnesium:Yes
- HEPES:No
- Sodium bicarbonate:Yes
- Sodium pyruvate:Yes
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:High
- Media Base:DMEM/F12 - Dulbecco's Modified Eagle Medium/F12 Nutrient Mixture