HBSS 1X-Hanks' Balanced Salt Solutions
New Product
HBSS 1X Hanks' Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions. HBSS is suitable for use in sealed culture flasks incubated in air.
- Calcium and magnesium salts
- Phenol red
- Sodium bicarbonate
: Store at 15 to 30 °C and protect from light. Shelf life: 24 months from date of manufacture.
- Calcium:Yes
- Sodium bicarbonate:Yes
- Phenol red:Yes
Frequently Bought Together