Order Entry
Canada
ContactUsLinkComponent
Anti-Interferon regulatory factor 9/IRF-9 Rabbit Monoclonal Antibody [clone: EPR24260-55]
Anti-Interferon regulatory factor 9/IRF-9 Rabbit Monoclonal Antibody [clone: EPR24260-55]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Interferon regulatory factor 9/IRF-9 Rabbit Monoclonal Antibody [clone: EPR24260-55]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB271043-100U
  • ABCA_AB271043-20UL
  • ABCA_AB271043-1ML
  • Antigen symbol:
  • Interferon regulatory factor 9/IRF-9
  • Interferon regulatory factor 9/IRF-9
  • Interferon regulatory factor 9/IRF-9
  • Antigen name:
  • ISGF3G
  • ISGF3G
  • ISGF3G
  • Antigen synonyms:
  • IRF9|Interferon regulatory factor 9|IRF-9|IFN-alpha-responsive transcription factor subunit|ISGF3 p48 subunit|Interferon-stimulated gene factor 3 gamma|Transcriptional regulator ISGF3 subunit gamma|ISGF-3 gamma
  • IRF9|Interferon regulatory factor 9|IRF-9|IFN-alpha-responsive transcription factor subunit|ISGF3 p48 subunit|Interferon-stimulated gene factor 3 gamma|Transcriptional regulator ISGF3 subunit gamma|ISGF-3 gamma
  • IRF9|Interferon regulatory factor 9|IRF-9|IFN-alpha-responsive transcription factor subunit|ISGF3 p48 subunit|Interferon-stimulated gene factor 3 gamma|Transcriptional regulator ISGF3 subunit gamma|ISGF-3 gamma
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR24260-55
  • EPR24260-55
  • EPR24260-55
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular weight:
  • 43696 Da
  • 43696 Da
  • 43696 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage temperature:
  • Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
  • Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
  • Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
  • Shipping temperature:
  • +4 °C
  • +4 °C
  • +4 °C
  • Tested applications:
  • ICC/IF, IHC-P
  • ICC/IF, IHC-P
  • ICC/IF, IHC-P

Specifications

Frequently Bought Together